Learn More
Crescent Chemical Co Inc PEPSIN PORCINE CA. 15 MILLIANS
SDP
Catalog No. 50247991
Encompass_Preferred
Pepsin porcine ca. 15 milliAnson units/mg 2xcryst. lyophil. For the degradation of proteins. Pepsin preferentially hydrolyzes those peptide linkages which involve the amino group contributed by the aromatic amino acids phenylalanine, tyrosine and tryptophan. Size - 5G Storage Condi
Catalog No. 50-247-991 Supplier Crescent Chemical Co Inc Supplier No. SE31820.02
Pepsin porcine ca. 15 milliAnson units/mg 2xcryst. lyophil. For the degradation of proteins. Pepsin preferentially hydrolyzes those peptide linkages which involve the amino group contributed by the aromatic amino acids phenylalanine, tyrosine and tryptophan. Size - 5G Storage Condi