Learn More
Crescent Chemical Co Inc PEPSIN PORCINE CA. 15 MILLIANS
SDP
Supplier:  Crescent Chemical Co Inc SE31820.01
Encompass_Preferred
Pepsin porcine ca. 15 milliAnson units/mg 2xcryst. lyophil. For the degradation of proteins. Pepsin preferentially hydrolyzes those peptide linkages which involve the amino group contributed by the aromatic amino acids phenylalanine, tyrosine and tryptophan. Size - 1G Storage Condi
Catalog No. 50-247-990