Learn More
Sigma Aldrich Fine Chemicals Biosciences Pepsin from porcine gastric mucosa, 9001-75-6, MFCD00081840, 25g
Catalog No. NC1570093
Encompass
Powder, ≥250 units/mg solid, Synonym: Pepsin A, Pepsin from hog stomach; Pepsin is a peptidase used to digest proteins and is commonly used in the preparation of Fab fragments from antibodies.
Catalog No. NC1570093 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P700025G
Powder, ≥250 units/mg solid, Synonym: Pepsin A, Pepsin from hog stomach; Pepsin is a peptidase used to digest proteins and is commonly used in the preparation of Fab fragments from antibodies.