Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Pepsin, porcine stomach

Catalog No. AAJ6167909
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6167909 10 g
AAJ6167906 5 g
AAJ6167903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6167909 Supplier Thermo Scientific Chemicals Supplier No. J6167909
Add to Cart

Pepsin, CAS # 9001-75-6, is a peptidase isolated from porcine gastric mucosa and used frequently to prepare Fab fragments from antibodies. | CAS: 9001-75-6

Pepsin is commonly used in the preparation of Fab fragments from antibodies. Pepsin, from porcine gastric mucosa, has been used to hydrolyze dry cervical samples in mice. has been used in the digestion of crude wheat gliadin. It has been used along with other enzymes to demonstrate the effects of fixation and enzymatic digestion in immunohistochemical assays, using paraffin embedded tissue. It has been used for digestion (before using immunoperoxidase techniques) to reduce non-specific background staining in sections of bronchial tissues. The enzyme has also been used in the preparation of F(ab)2 fragment from IgG.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Pepsin hydrolyzes peptide bonds, but does not have the ability to hydrolyze amide or ester linkages. This compound cleaves peptides with an aromatic acid on either side of the peptide bond

Application

  • Pepsin from the porcine stomach is used to form Fab fragments from antibodies and prepare F(ab)2 fragments from IgG
  • It is applied to hydrolyze dry cervical samples in mice
  • In biochemical studies, it has been used to demonstrate the effects of fixation and enzymatic digestion in immunohistochemical assays using paraffin-embedded tissue
  • It is also used for digestion (before using immunoperoxidase techniques) to reduce non-specific background staining in sections of bronchial tissues
  • This compound catalyzes the hydrolysis of aminoacyl-proline to proline
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9001-75-6
Synonym EC 3.4.23.1; Pepsin A

Specifications

Content And Storage -30°C to -10°C
Form Powder
Quantity 10 g
Solubility Dilute Aqueous Buffers, Water
Product Type Pepsin
EINECSNumber 232-629-3
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.