Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Pepsin from Porcine Stomach

Catalog No. AAJ20015A1 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ20015A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ20015A1 Supplier Thermo Scientific Chemicals Supplier No. J20015A1
Add to Cart

missing translation for 'validMainframeMsg'

Derived from Porcine stomach

  • One unit causes an increase in absorbance at 280nm of 0.001/min. at 37°C at pH 2.0.
TRUSTED_SUSTAINABILITY

Specifications

CAS 9001-75-6
Form Powder
Quantity 1 kg
Synonym EC 3.4.23.1,Pepsin A
Product Type Pepsin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.