Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Pepsin Powder
SCI_ED

Catalog No. S25459A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
30 g
100 g
Packaging:
Powder
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25459A 100 g Powder
S25459 30 g Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25459A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

White to yellow pepsin powder. Ideal for digestive enzyme experiments in biology and chemistry classrooms.

  • Material: Pepsin Powder
  • Color: White to yellow
  • Quantity: 100 grams
  • Packaging: Powder form
  • Usage: For educational use only – no Certificate of Analysis (C of A) available
  • Ideal For: Enzyme activity studies, protein digestion experiments, biology and chemistry classrooms

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 9001-75-6

Specifications

Color White to Yellow
Physical Form Flakes or Powder
Quantity 100 g
Packaging Powder
Chemical Name or Material Pepsin Powder
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.