Learn More
Pepsin Reagent, Ready to Use, Antigen Retriever, Sigma-Aldrich
Catalog No. 01258906 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
01-258-906 100 mL
1 options
Catalog No. 01-258-906 Supplier MilliporeSigma Supelco Supplier No. R2283100ML
Add to Cart
Add to Cart
Pepsin is used for proteolytic digestion of formalin-fixed paraffin-embedded (FFPE) tissue sections prior to the application of antibodies in immunohistochemistry (IHC). It has been used to enhance the permeabilization of fungal cell walls during the fluorescence in situ hybridization (FISH) process and utilized during the RNA scope in situ hybridization process to perform antigen retrieval.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Pepsin
Quantity 100 mL
Solution Type Buffer
Form Liquid