Learn More
Biomatik Corporation Pepsinogen A (Pga) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29499
Encompass_Preferred
Pepsinogen A (Pga) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Pepsinogen A (PGA). The Pepsinogen A (Pga) Monoclonal Antibody has been validated for the following applications: WB, IHC. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Val63~Phe163 + Gly206~Ser295. Applications: WB, IHC. Target Synonyms: Pepsinogen A (PGA). Accession Number: P00790. Gene ID: 643834. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4829