Learn More
Medchemexpress LLC HY-P0018 10mg , Pepstatin CAS:26305-03-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP001810MG
Encompass_Preferred
HY-P0018 10mg Pepstatin CAS: 26305-03-3 Pepstatin (Pepstatin A) is a specific aspartic protease inhibitor produced by actinomycetes, with IC50s of 4.5 nM, 6.2 nM, 150 nM, 290 nM, 520 nM and 260 nM for hemoglobin-pepsin, hemoglobin-proctase, casein-pepsin, casein-proctase, casein-acid protease and hemoglobin-acid protease, respectively. Pepstatin Ammonium also inhibits HIV protease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9836