Learn More
Research Products International Corp Pepstatin A, 100 Milligrams
SDP
Catalog No. 501368504
Encompass_Preferred
An inhibitor of aspartic proteases. Forms a 1:1 complex with proteases such as pepsin, renin and cathepsin D. Highly selective and does not inhibit thiol proteases, neutral proteases, or serine proteases.
Catalog No. 50-136-8504 Supplier Research Products International Corp Supplier No. P301000.1
An inhibitor of aspartic proteases. Forms a 1:1 complex with proteases such as pepsin, renin and cathepsin D. Highly selective and does not inhibit thiol proteases, neutral proteases, or serine proteases.