Learn More
Alkali Scientific Pepstatin A, 100 Mg
SDP
Catalog No. 5000661316
Encompass_Preferred
Pepstatin A is a peptide-based protease inhibitor used in a variety of laboratory applications, including protein stabilization, enzyme function studies, and protease inhibition assays. This reagent inhibits aspartic proteases, which play an important role in protein degradation pathways. Available in 100 mg quantities, Pepstatin A is especially useful in experiments where the preservation of protein integrity is essential, such as protein isolation and purification, enzyme assays, and cellular studies. By preventing proteolytic cleavage, Pepstatin A helps to ensure that the proteins under study remain in their active forms, making it a vital reagent for researchers working in molecular biology and biochemistry.
Catalog No. 50-006-61316 Supplier Alkali Scientific Supplier No. PEP004CP
Pepstatin A is a peptide-based protease inhibitor used in a variety of laboratory applications, including protein stabilization, enzyme function studies, and protease inhibition assays. This reagent inhibits aspartic proteases, which play an important role in protein degradation pathways. Available in 100 mg quantities, Pepstatin A is especially useful in experiments where the preservation of protein integrity is essential, such as protein isolation and purification, enzyme assays, and cellular studies. By preventing proteolytic cleavage, Pepstatin A helps to ensure that the proteins under study remain in their active forms, making it a vital reagent for researchers working in molecular biology and biochemistry.