Learn More
Bachem Pepstatin A
SDP
Supplier:  Bachem 4031136.0100
Encompass_Preferred
Pepstatin A, Isovaleryl-Val-Val-Sta-Ala-Sta-OH, 26305-03-3, C34H63N5O9, Mr 685.90, 100mg. Pepstatin A (isovaleryl-pepstatin), a low molecular weight inhibitor of acid proteases, e.g. pepsin, renin, cathepsin D, and plasmepsin II. For the Plasmodium falciparum aspartyl protease plasmepsin II, Hill et al determined an IC50 of 0.1 nM.
Catalog No. 50-260-523