Learn More
Selleck Chemical LLC Pepstatin A 10mg 26305-03-3 Pepstatin
SDP
Catalog No. 501364722
Encompass_Preferred
Pepstatin A (Pepstatin) is a potent aspartic protease inhibitor, and also inhibits HIV replication. Pepstatin A is also an inhibitor of cathepsins D and cathepsins E. Pepstatin A inhibits autophagy by suppressing lysosomal proteases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4722 Supplier Selleck Chemical LLC Supplier No. S738110MG
Pepstatin A (Pepstatin) is a potent aspartic protease inhibitor, and also inhibits HIV replication. Pepstatin A is also an inhibitor of cathepsins D and cathepsins E. Pepstatin A inhibits autophagy by suppressing lysosomal proteases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.