Learn More
Ubpbio An irreversible inhibitor of aspartic acid proteases. CAS# 26305-03-3, > 98% purity
Catalog No. 502339331
Encompass_Preferred
Pepstatin A is an irreversible inhibitor of aspartic acid proteases, including pepsin, cathepsin D, cathepsin E, chymosin, and HIV proteases. It inhibits lysosomal proteases and autophagy. Typical working concentration is 1 µM.
Catalog No. 50-233-9331 Supplier Ubpbio Supplier No. P105410
Pepstatin A is an irreversible inhibitor of aspartic acid proteases, including pepsin, cathepsin D, cathepsin E, chymosin, and HIV proteases. It inhibits lysosomal proteases and autophagy. Typical working concentration is 1 µM.