Learn More
Apexbio Technology LLC Aspartic proteinases inhibitor 26305-03-3. MFCD00060740
SDP
Catalog No. 501010463
Encompass_Preferred
Pepstatin A is a well-known inhibitor of aspartic proteinases with IC50 values of 15 μM, 2 μM, <5 nM and <40 nM for human renin, HIV protease, pepsin and cathepsin D, respectively. Pepstatin A is a pentapeptide. It was originally isolated from the microbe. As a most potent renin inhibitor, pepstatinA inhibited porcine renin and human renin at weak acid pH value with IC50 values of 0.32 and 15 μM, respectively. The disadvantage is that, pepstatin Ais hydrophobic. To couple the charged hydrophilic residues to the C-terminal of pepstatinA can increase its solubility. Besides the renin, pepstatin A was also reported to have inhibitory effects on HIV protease and subsequently suppressed the virus replication. In cultured H9 cells, pepstatinA treatment blocked the proteolytic processing of the virus gag precursor and inhibited the production of infectious HIV. Other sizes are available. Please inqury us for quote.
Catalog No. 50-101-0463 Supplier Apexbio Technology LLC Supplier No. A257110
Pepstatin A is a well-known inhibitor of aspartic proteinases with IC50 values of 15 μM, 2 μM, <5 nM and <40 nM for human renin, HIV protease, pepsin and cathepsin D, respectively. Pepstatin A is a pentapeptide. It was originally isolated from the microbe. As a most potent renin inhibitor, pepstatinA inhibited porcine renin and human renin at weak acid pH value with IC50 values of 0.32 and 15 μM, respectively. The disadvantage is that, pepstatin Ais hydrophobic. To couple the charged hydrophilic residues to the C-terminal of pepstatinA can increase its solubility. Besides the renin, pepstatin A was also reported to have inhibitory effects on HIV protease and subsequently suppressed the virus replication. In cultured H9 cells, pepstatinA treatment blocked the proteolytic processing of the virus gag precursor and inhibited the production of infectious HIV. Other sizes are available. Please inqury us for quote.