Learn More
Apexbio Technology LLC Pepstatin A 26305-03-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063554
Encompass_Preferred
Pepstatin A is a pentapeptide functioning as an inhibitor targeting aspartic proteases including pepsin renin HIV protease and cathepsin D It inhibits human renin and HIV protease with IC50 values of approximately 15 M and 2 M respectively and shows inhibitory activity toward pepsin and cathepsin D at IC50 values below 5 M and 40 M Mechanistically Pepstatin A acts through binding to the catalytic site of aspartic proteases restricting proteolytic activity In biomedical research it is utilized to study viral protein processing osteoclast differentiation mediated by cathepsins and as a standard tool in enzyme inhibition assays examining aspartic protease function
Catalog No. 50-000-63554 Supplier Apexbio Technology LLC Supplier No. A25715.1
Pepstatin A is a pentapeptide functioning as an inhibitor targeting aspartic proteases including pepsin renin HIV protease and cathepsin D It inhibits human renin and HIV protease with IC50 values of approximately 15 M and 2 M respectively and shows inhibitory activity toward pepsin and cathepsin D at IC50 values below 5 M and 40 M Mechanistically Pepstatin A acts through binding to the catalytic site of aspartic proteases restricting proteolytic activity In biomedical research it is utilized to study viral protein processing osteoclast differentiation mediated by cathepsins and as a standard tool in enzyme inhibition assays examining aspartic protease function