Learn More
Research Products International Corp PEPSTATIN A, 25 MG
SDP
Catalog No. 50488907
Encompass_Preferred
An inhibitor of aspartic proteases. Forms a 1:1 complex with proteases such as pepsin, renin and cathepsin D. Highly selective and does not inhibit thiol proteases, neutral proteases, or serine proteases.
Catalog No. 50-488-907 Supplier Research Products International Corp Supplier No. P301000.025
An inhibitor of aspartic proteases. Forms a 1:1 complex with proteases such as pepsin, renin and cathepsin D. Highly selective and does not inhibit thiol proteases, neutral proteases, or serine proteases.