Learn More
Research Products International Corp Pepstatin A, 250 MG
SDP
Supplier:  Research Products International Corp P301000.25
Encompass_Preferred
An inhibitor of aspartic proteases. Forms a 1:1 complex with proteases such as pepsin, renin and cathepsin D. Highly selective and does not inhibit thiol proteases, neutral proteases, or serine proteases.
Catalog No. 50-197-6901