Learn More
Crescent Chemical Co Inc PEPSTATIN A
SDP
Supplier:  Crescent Chemical Co Inc SE52682.03
Encompass_Preferred
Pepstatin A Inhibitor of aspartic proteases, e.g. pepsin, renin, cathepsin D (1 - 3) and of retroviral proteases (4 - 7). Size - 25MG Storage Conditions - +2 °C TO +8 °C Catalog Number - 52682.03 CAS 26305-03-3
Catalog No. 50-249-206