Learn More
Alkali Scientific Pepstatin A, 5 Mg
SDP
Catalog No. 5000661214
Encompass_Preferred
Pepstatin A is a potent peptide-based protease inhibitor used for stabilizing proteins by inhibiting aspartic proteases. This inhibitor is essential for research studies that focus on protein function, structure, and enzymatic activity, particularly in the fields of molecular biology, biochemistry, and pharmacology. Pepstatin A has been used in research to investigate processes like protein degradation and intracellular signaling. By blocking protease activity, it helps to preserve protein integrity during experimental procedures, making it an invaluable reagent for studying cellular processes, enzyme kinetics, and metabolic pathways in both isolated and cellular systems.
Catalog No. 50-006-61214 Supplier Alkali Scientific Supplier No. PEP001CP
Add to Cart
Add to Cart
Pepstatin A is a potent peptide-based protease inhibitor used for stabilizing proteins by inhibiting aspartic proteases. This inhibitor is essential for research studies that focus on protein function, structure, and enzymatic activity, particularly in the fields of molecular biology, biochemistry, and pharmacology. Pepstatin A has been used in research to investigate processes like protein degradation and intracellular signaling. By blocking protease activity, it helps to preserve protein integrity during experimental procedures, making it an invaluable reagent for studying cellular processes, enzyme kinetics, and metabolic pathways in both isolated and cellular systems.