Learn More
Carbosynth LLC. Pepstatin A (Purity Higher than 90% by HPLC), 100mg, MW: 685.89, Molecular formula: C34H63N5O9, CAS# 26305-03-3
SDP
Supplier:  Carbosynth LLC. IPA4397100MG
Encompass_Preferred
Synonyms: Isovaleryl-L-Valyl-L-Valyl[(3S,4S)-4-Amino-3-Hydroxy-6-Methylheptanoyl]-L-Alanyl [(3S,4S)-4-Amino-3-Hydroxy-6-Methylheptanoic Acid], Purity: , higher than 90% (HPLC), Application: Microbial Product Inhibitor for Pepsin, Cathepsin D/E and Renin, Storage Temperature: -20°C, References: H. Umezawa, T. Aoyagi, H. Morishima, M. Matsuzaki, M. Hamada, and T. Takeuchi, J. Antibiotics, 23, 259 (1970). (Original), T. Aoyagi, H. Morishima, R. Nishizawa, S. Kunimoto, T. Takeuchi, H. Umezawa, and H. Ikezawa, J. Antibiotics, 25, 689 (1972). (Biological Activity)
Catalog No. 50-168-5889