Learn More
Selleck Chemical LLC Pepstatin A
SDP
Catalog No. 5000004373
Encompass_Preferred
Pepstatin A (Pepstatin) is a potent aspartic protease inhibitor and also inhibits HIV replication Pepstatin A is also an inhibitor of cathepsins D and cathepsins E Pepstatin A inhibits autophagy by suppressing lysosomal proteases
Catalog No. 50-000-04373 Supplier Selleck Chemical LLC Supplier No. S738150MG
Pepstatin A (Pepstatin) is a potent aspartic protease inhibitor and also inhibits HIV replication Pepstatin A is also an inhibitor of cathepsins D and cathepsins E Pepstatin A inhibits autophagy by suppressing lysosomal proteases