Learn More
Selleck Chemical LLC Pepstatin A
Supplier: Selleck Chemical LLC S73811G
Pepstatin A (Pepstatin) is a potent aspartic protease inhibitor and also inhibits HIV replication Pepstatin A is also an inhibitor of cathepsins D and cathepsins E Pepstatin A inhibits autophagy by suppressing lysosomal proteases