Learn More
AdipoGen Pepstatin A methyl ester
SDP
Catalog No. 502562412
Encompass_Preferred
Chemical. CAS 29169-91-3. Formula C35H65N5O9. MW 699.9. Cell permeable methyl ester derivative of Pepstatin A. Potent, non-competitive, transition-state analog inhibitor of gamma-secretase.
Catalog No. 50-256-2412 Supplier AdipoGen Supplier No. AGCP37004M001
Chemical. CAS 29169-91-3. Formula C35H65N5O9. MW 699.9. Cell permeable methyl ester derivative of Pepstatin A. Potent, non-competitive, transition-state analog inhibitor of gamma-secretase.