Learn More
Research Products International Corp PEPSTATIN A, 5 MG
SDP
Catalog No. 50550428
Encompass_Preferred
An inhibitor of aspartic proteases. Forms a 1:1 complex with proteases such as pepsin, renin and cathepsin D. Highly selective and does not inhibit thiol proteases, neutral proteases, or serine proteases.
Catalog No. 50-550-428 Supplier Research Products International Corp Supplier No. P301000.005
An inhibitor of aspartic proteases. Forms a 1:1 complex with proteases such as pepsin, renin and cathepsin D. Highly selective and does not inhibit thiol proteases, neutral proteases, or serine proteases.