Learn More
Enzo Life Sciences Pepstatin A (synthetic) (100mg). CAS: 26305-03-3
SDP
Catalog No. 502008305
Encompass_Preferred
Protease inhibitor. Inhibitor of aspartate (acid) proteases, including pepsin, cathepsin D, renin, chymosin, bacterial aspartic proteinases and HIV proteases. Efficiently inhibits Fcγ receptor induction by IFN-γ. Blocks the endocytic pathway followed by an antibody when it enters the cell at the level of early endosomal compartments, resulting in inhibition of autoantigen presentation or viral transmission. Suppresses p-53-dependent apoptosis in lymphoid cells as well as TNFα-induced apoptosis in U937 cells. Inhibits degradation of autophagic cargo inside autophagolysosomes. Alternative name: Iva-Val-Val-Sta-Ala-Sta, i-Valeryl-L-Val-L-Val-AHMHA-L-Ala-AHMHA. Formula: C34H63N5O9. MW: 685.9. Purity: ≥98% (HPLC). Appearance: White to off-white solid. MI: 14: 7147. Solubility: Soluble in DMSO (10mg/ml), methanol (1mg/ml) or acetic acid. Long Term Storage: -20°C. Use/Stability: Stable for at least 2 years after receipt when stored at -20°C. Handling: Protect from light.
Catalog No. 50-200-8305 Supplier Enzo Life Sciences Supplier No. ALX260085M100
Protease inhibitor. Inhibitor of aspartate (acid) proteases, including pepsin, cathepsin D, renin, chymosin, bacterial aspartic proteinases and HIV proteases. Efficiently inhibits Fcγ receptor induction by IFN-γ. Blocks the endocytic pathway followed by an antibody when it enters the cell at the level of early endosomal compartments, resulting in inhibition of autoantigen presentation or viral transmission. Suppresses p-53-dependent apoptosis in lymphoid cells as well as TNFα-induced apoptosis in U937 cells. Inhibits degradation of autophagic cargo inside autophagolysosomes. Alternative name: Iva-Val-Val-Sta-Ala-Sta, i-Valeryl-L-Val-L-Val-AHMHA-L-Ala-AHMHA. Formula: C34H63N5O9. MW: 685.9. Purity: ≥98% (HPLC). Appearance: White to off-white solid. MI: 14: 7147. Solubility: Soluble in DMSO (10mg/ml), methanol (1mg/ml) or acetic acid. Long Term Storage: -20°C. Use/Stability: Stable for at least 2 years after receipt when stored at -20°C. Handling: Protect from light.