Learn More
Medchemexpress LLC HY-P0018B 10mg Medchemexpress, Pepstatin Ammonium CAS: Purity:>98%
SDP
Catalog No. 501933992
Encompass_Preferred
Medchemexpress, HY-P0018B 10mg Pepstatin Ammonium CAS: Pepstatin Ammonium is a specific aspartic protease inhibitor produced by actinomycetes, with IC 50 s of 4.5 nM, 6.2 nM, 150 nM, 290 nM, 520 nM and 260 nM for hemoglobin-pepsin, hemoglobin-proctase, casein-pepsin, casein-proctase, casein-acid protease and hemoglobin-acid protease, respectively. Pepstatin Ammonium also inhibits HIV protease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3992 Supplier Medchemexpress LLC Supplier No. HYP0018B10MG
Medchemexpress, HY-P0018B 10mg Pepstatin Ammonium CAS: Pepstatin Ammonium is a specific aspartic protease inhibitor produced by actinomycetes, with IC 50 s of 4.5 nM, 6.2 nM, 150 nM, 290 nM, 520 nM and 260 nM for hemoglobin-pepsin, hemoglobin-proctase, casein-pepsin, casein-proctase, casein-acid protease and hemoglobin-acid protease, respectively. Pepstatin Ammonium also inhibits HIV protease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.