Learn More
Medchemexpress LLC Pepstatin Trifluoroacetate | 98.8% | 799.92 | 5 MG
SDP
Catalog No. 5000426234
Encompass_Preferred
Pepstatin Trifluoroacetate is a specific, orally active aspartic protease inhibitor derived from actinomycetes. It effectively inhibits various enzymes such as hemoglobin-pepsin, hemoglobin-proctase, casein-pepsin, casein-proctase, casein-acid protease, and hemoglobin-acid protease with IC50 values ranging from 4.5 nM to 520 nM. This compound also demonstrates inhibition of HIV protease.
  • Specific, orally active aspartic protease inhibitor
  • Inhibits hemoglobin-pepsin with an IC50 of 4.5 nM
  • Inhibits HIV protease
  • Demonstrates low toxicity in animal studies
  • Suppresses stomach ulceration in animal models
Catalog No. 50-004-26234 Supplier Medchemexpress LLC Supplier No. HYP0018A5MG
Pepstatin Trifluoroacetate is a specific, orally active aspartic protease inhibitor derived from actinomycetes. It effectively inhibits various enzymes such as hemoglobin-pepsin, hemoglobin-proctase, casein-pepsin, casein-proctase, casein-acid protease, and hemoglobin-acid protease with IC50 values ranging from 4.5 nM to 520 nM. This compound also demonstrates inhibition of HIV protease.
  • Specific, orally active aspartic protease inhibitor
  • Inhibits hemoglobin-pepsin with an IC50 of 4.5 nM
  • Inhibits HIV protease
  • Demonstrates low toxicity in animal studies
  • Suppresses stomach ulceration in animal models