Learn More
Discovery Life Sciences Human Peptide Transporter 1, ≥10 million cells
SDP
Supplier:  Discovery Life Sciences 354860
Encompass
Corning SLC TransportoCells products are high performance mammalian cells transiently overexpressing a single human SLC transporter protein. They are ideal cell-based models for studying transporter activities in reaction phenotyping, drug-drug interaction and drug clearance studies.
Catalog No. NC1347173