Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bruker Daltonics Peptide Calibration Standard II, 5 tubes, 700 to 3200 Da, nine standard peptides, for MALDI-TOF MS, 5 x 250 calibration spots

Catalog No. NC1162644
Encompass

The Bruker Peptide Calibration Standard II is specifically designed for precise calibration and performance verification of MALDI-TOF mass spectrometers. It contains a mixture of nine key peptides, optimized for accurate measurements in peptide analysis. This product ensures reliable results across a low mass range of 700 to 3200 Da, making it an essential tool for researchers.

  • Application for calibration and testing of MALDI-TOF MS
  • Compatible with MALDI-TOF mass spectrometers
  • Mass range of 700 to 3200 Da
  • Contains nine standard peptides
  • Package includes five tubes
  • Provides 5 x 250 calibration spots
  • Recommended matrix is α-Cyano-4-hydroxycinnamic acid
  • Shipping at ambient temperatures
  • Storage condition on arrival: less than 0 °C
  • Storage condition dissolved: aliquoted and frozen; do not refreeze
  • For research use only; not for diagnostic procedures

Catalog No. NC1162644 Supplier Bruker Daltonics Supplier No. 8222570
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Bruker Peptide Calibration Standard II is specifically designed for precise calibration and performance verification of MALDI-TOF mass spectrometers. It contains a mixture of nine key peptides, optimized for accurate measurements in peptide analysis. This product ensures reliable results across a low mass range of 700 to 3200 Da, making it an essential tool for researchers.

  • Application for calibration and testing of MALDI-TOF MS
  • Compatible with MALDI-TOF mass spectrometers
  • Mass range of 700 to 3200 Da
  • Contains nine standard peptides
  • Package includes five tubes
  • Provides 5 x 250 calibration spots
  • Recommended matrix is α-Cyano-4-hydroxycinnamic acid
  • Shipping at ambient temperatures
  • Storage condition on arrival: less than 0 °C
  • Storage condition dissolved: aliquoted and frozen; do not refreeze
  • For research use only; not for diagnostic procedures

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.