Learn More
Bruker Daltonics Peptide Calibration Standard, 5 tubes, 7 standard peptides, Mass Range 1000 to 3500 Da, Ambient shipping temperature, Less than 0 °C storage temperature
Supplier:  Bruker Daltonics 8206195
Encompass
Peptide Calibration Standard, 5 tubes, 7 standard peptides, Mass Range 1000 to 3500 Da, Ambient shipping temperature, Less than 0 °C storage temperature
Catalog No. NC9846988