Learn More
Waters Corp Peptide CSH C18 Column, 130 Å, 2.5 μm
Supplier:  Waters Corp 186009908/PEPTIDE
Encompass_Preferred
This column is optimized for peptide separations, utilizing particles with precisely defined positive charges to achieve excellent peak shape with FA- or TFA-containing mobile phases. It ensures enhanced reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Optimized for peptide separations.
  • Particles with low, defined positive charges yield excellent peak shape with FA- or TFA-containing mobile phases.
  • Quality controlled with cytochrome c digest standards for peptide analysis.
  • Minimal dependence on strong ion pairing agents for LC applications.
  • Enhances reproducibility, peak shape, and recovery for metal-sensitive analytes.
Catalog No. NC3851379