Learn More
Bioworld Peptide Pre-Cleaning Buffer, 10 mL
Supplier:  Bioworld 220400471
Encompass_Preferred
Peptide Precleaning Buffer, 10X For clearification of proteins to eliminate nucleic acids and polysaccharides before electrophoresis. Total protein preparation and cleaning solution Technical Information:For use with 1D gels Storage: -20C Sold as 10 - 1ml quantities of ready-to-use solution each
Catalog No. 50-253-047