Learn More
Genscript Corporation MDYKDHDGDYKDHDIDYKDDDDK peptide
Supplier:  Genscript Corporation RP2108710
Encompass
A fusion tag, called MDYKDHDGDYKDHDIDYKDDDDK including an enterokinase-cleavage site, was specifically designed for immunoaffinity chromatography.
Catalog No. NC2096762