Learn More
Bachem Peptide YY (3-36) (canine, mouse, porcine, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4044052.1000
Encompass_Preferred
Peptide YY (3-36) (canine, mouse, porcine, rat) Trifluoroacetate, H-Ala-Lys-Pro-Glu-Ala-Pro-Gly-Glu-Asp-Ala-Ser-Pro-Glu-Glu-Leu-Ser-Arg-Tyr-Tyr-Ala-Ser-Leu-Arg-His-Tyr-Leu-Asn-Leu-Val-Thr-Arg-Gln-Arg-Tyr-NH2, 126339-09-1, C176H272N52O54, Mr 3980.41, 1mg. PYY (3-36) (canine, mouse, porcine, rat). Peptide YY (3-36) and peptide YY are both synthesized by the gastrointestinal tract and released into the circulation after a meal. Peptide YY (3-36) is a Y2 receptor subtype agonist, whereas peptide YY is non-selective for Y1 and Y2 receptor subtypes. It has been suggested that Y1 and Y2 receptor subtype binding affinities depend on their secondary and tertiary solution state structures.
Catalog No. 50-260-020