Learn More
Enzo Life Sciences Peptide YY (3-36) monoclonal antibody (4) (1 mg)
SDP
Supplier:  Enzo Life Sciences BPDABS051041
Encompass_Preferred
Clone: 4. Isotype: IgG2aκ. Immunogen: Synthetic peptide corresponding to aa 3-26 of human Peptide YY coupled to carrier. Species reactivity: Human. Applications: ELISA, WB. Purity: Protein A/G-affinity purified. Formulation: Liquid. In 0.01M phosphate buffer, pH 7.4, containing 0.5M sodium chloride and 15mM sodium azide.
Catalog No. 50-201-2433