Learn More
Enzo Life Sciences Peptide YY (human) ELISA Kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ENZKIT1330001
Encompass_Preferred
The Peptide YY ELISA kit is a colorimetric immunometric immunoassay kit with results in 3.5 hrs. Alternative name: Peptide tyrosine tyrosine, PPY, Pancreatic peptide YY. Assay Time: 3.5 hrs. Applications: ELISA, Colorimetric detection . Species reactivity: Human .
Catalog No. 50-201-3828