Learn More
Eurofins Discoverx Peptide YY, Human, 0.5 mg. (92-1063)
Supplier:  Eurofins Discoverx 921063
Encompass
Pancreatic peptide tyrosine tyrosine (peptide YY or PYY) is a 36 amino acid peptide released in the ileum and colon in response to feeding. It acts through neuropeptide Y (NPY) receptors to reduce appetite, inhibit gastric motility, and increase water and electrolyte absorption in the colon. It is a selective agonist for NPY2R over NPY1R (0.11 nM and 1050 nM, respectively). Provided as 500 μg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9604