Learn More
Enzo Life Sciences Peptide tyrosine tyrosine polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences BMLPA12540100
Encompass_Preferred
Immunogen: Natural pig PYY (peptide with tyrosine; HYPAKPEAPGEDASPEELSRYYASLRHYLNLVTRQRY-NH2.). Source: Whole rabbit serum. Species reactivity: Species independent. Specificity: Recognizes PYY in a wide range of species. Applications: IHC, WB. Formulation: Liquid. Antiserum containing 10mM sodium azide.
Catalog No. 50-201-1369