Learn More
Medchemexpress LLC HY-P1021A 5mg Medchemexpress, Peptide YY (PYY) (3-36), human (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1021A5MG
Encompass_Preferred
Medchemexpress, HY-P1021A 5mg Peptide YY (PYY) (3-36), human (TFA) CAS: Peptide YY (PYY) (3-36), human (TFA) is a gut hormone peptide that acts as a Y2 receptor agonist to reduce appetite [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4130