Learn More
Medchemexpress LLC Peptide YY (PYY), human | 118997-30-1 | 99.2% | 4309.75 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP15141MG
Encompass_Preferred
Peptide YY (PYY) is a gut hormone that regulates appetite and inhibits pancreatic secretion. It acts through Neuropeptide Y receptors and belongs to the pancreatic polypeptide family. This 36-amino acid peptide is secreted from L cells in the gastrointestinal tract and can strongly inhibit secretin-stimulated pancreatic secretion, reducing bicarbonate and fluid secretion. It is also found in blood vessels, causing vasoconstriction.
  • Regulates appetite and inhibits pancreatic secretion.
  • Mediates effects through Neuropeptide Y receptors.
  • Composed of 36 amino acids.
  • Secreted from neuroendocrine cells in the gastrointestinal tract.
  • Strongly inhibits secretin-stimulated pancreatic secretion.
  • Decreases pancreatic bicarbonate and fluid secretion.
  • Causes vasoconstriction of the vascular tree.
  • Solid, white to off-white appearance.
Catalog No. 50-004-22227