Learn More
Waters Corp PeptideWorks Tryptic Protein Digestion Refill Kit, without Unfolding, 96 Analysis, MS Grade
Catalog No. 502391064 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This refill kit provides desalting cartridges and tryptic enzymatic digestion with RapiZyme Trypsin. It is designed for customers using their own unfolding reagents, enabling automatable, high-efficiency, reproducible peptide maps in under 2.5 hours for 4 to 96 samples.
  • Processes up to 96 analyses in a sample preparation workflow.
  • Compatible with UPLC/UHPLC/HPLC separation and MS, optical, & UV detection for LC-MS systems.
  • Mass Spectrometry (MS) grade trypsin ensures quick, clean, & complete digestions in 30 minutes.
  • Digestion speed is under 2.5 hours, 4x faster than average home-brew methods.
  • Achieves 78% reduction in missed cleavages & 98% reduction in contaminating autolysis peaks.
  • Supports manual or automated processing.
  • Includes 96 desalting cartridges, 2 vials of 1 M CaCl2 salt concentrate, & 4 vials of trypsin.
Catalog No. 50-239-1064 Supplier Waters Corp Supplier No. 176005309
This refill kit provides desalting cartridges and tryptic enzymatic digestion with RapiZyme Trypsin. It is designed for customers using their own unfolding reagents, enabling automatable, high-efficiency, reproducible peptide maps in under 2.5 hours for 4 to 96 samples.
  • Processes up to 96 analyses in a sample preparation workflow.
  • Compatible with UPLC/UHPLC/HPLC separation and MS, optical, & UV detection for LC-MS systems.
  • Mass Spectrometry (MS) grade trypsin ensures quick, clean, & complete digestions in 30 minutes.
  • Digestion speed is under 2.5 hours, 4x faster than average home-brew methods.
  • Achieves 78% reduction in missed cleavages & 98% reduction in contaminating autolysis peaks.
  • Supports manual or automated processing.
  • Includes 96 desalting cartridges, 2 vials of 1 M CaCl2 salt concentrate, & 4 vials of trypsin.