Learn More
Waters Corp PeptideWorks Refill Kit, Tryptic Protein Digestion, Automatable, RapiZyme Trypsin
Catalog No. 502392460 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This refill kit provides components for automatable, high-efficiency, and reproducible peptide mapping, achieving results four times faster than average home-brew methods. It supports both manual digestion and automated processes.
  • Achieves automatable, high-efficiency, and reproducible peptide maps.
  • Includes Mass Spectrometry (MS) grade RapiZyme Trypsin for rapid, clean, and complete digestions in 30 minutes without autolysis peaks.
  • Contains 4 vials of RapiZyme Trypsin.
  • Components for denaturation (Guanidine Hydrochloride), reduction (Dithiothreitol), and alkylation (Iodoacetamide) are provided.
  • Supplied with Sep-Pak SEC Desalting Cartridges and 1M CaCl2 Salt Concentrate.
  • Supports up to 96 protein digestion analyses.
Catalog No. 50-239-2460 Supplier Waters Corp Supplier No. 176005312
This refill kit provides components for automatable, high-efficiency, and reproducible peptide mapping, achieving results four times faster than average home-brew methods. It supports both manual digestion and automated processes.
  • Achieves automatable, high-efficiency, and reproducible peptide maps.
  • Includes Mass Spectrometry (MS) grade RapiZyme Trypsin for rapid, clean, and complete digestions in 30 minutes without autolysis peaks.
  • Contains 4 vials of RapiZyme Trypsin.
  • Components for denaturation (Guanidine Hydrochloride), reduction (Dithiothreitol), and alkylation (Iodoacetamide) are provided.
  • Supplied with Sep-Pak SEC Desalting Cartridges and 1M CaCl2 Salt Concentrate.
  • Supports up to 96 protein digestion analyses.