Learn More
Waters Corp PeptideWorks Tryptic Protein Digestion Kit, Unfolding-Free, Automation-Ready, 2.5 hr
Catalog No. 502391380 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit provides all necessary reagents for protein digestion in peptide mapping applications, suitable for users preferring their own denaturation, reduction, and alkylation reagents. It offers a streamlined, automation-ready workflow, completing in under 2.5 hours, ensuring high-quality and reproducible protein digestion.
  • Streamlined and robust protein digestion workflow.
  • Flexible workflow: manual or automated, completes in under 2.5 hours.
  • High-quality digestion with recombinant trypsin.
  • Reproducible peptide patterns for biopharmaceutical protein characterization.
  • Rapid digestion: 4x faster than average home-brew methods.
  • Improved digestion quality: reduced missed cleavages and contaminating autolysis peaks.
  • Optimized performance: lyophilized trypsin with LC-MS compatible additive.
Catalog No. 50-239-1380 Supplier Waters Corp Supplier No. 176005308
This kit provides all necessary reagents for protein digestion in peptide mapping applications, suitable for users preferring their own denaturation, reduction, and alkylation reagents. It offers a streamlined, automation-ready workflow, completing in under 2.5 hours, ensuring high-quality and reproducible protein digestion.
  • Streamlined and robust protein digestion workflow.
  • Flexible workflow: manual or automated, completes in under 2.5 hours.
  • High-quality digestion with recombinant trypsin.
  • Reproducible peptide patterns for biopharmaceutical protein characterization.
  • Rapid digestion: 4x faster than average home-brew methods.
  • Improved digestion quality: reduced missed cleavages and contaminating autolysis peaks.
  • Optimized performance: lyophilized trypsin with LC-MS compatible additive.