Learn More
TOKU-E Blasticidin S Hydrochloride, 3513-03-9, 100 mg
Catalog No. 502551833
Encompass_Preferred
peptidyl nucleoside antibiotic, C17H26N8O5 HCl, selection antibiotic, Inhibits protein synthesis. Useful in cell culture, microbiology.
Catalog No. 50-255-1833 Supplier TOKU-E Supplier No. B001100MG
peptidyl nucleoside antibiotic, C17H26N8O5 HCl, selection antibiotic, Inhibits protein synthesis. Useful in cell culture, microbiology.