Learn More
Research Products International Corp Peptone, 1 Gram Capsules, 1 Kilogram
SDP
Catalog No. 50488896
Encompass_Preferred
Each capsule contains 1g Peptone. Convenient and easy to use - simply add required number of premeasured capsules according to formulation being prepared without weighing and measuring.
Catalog No. 50-488-896 Supplier Research Products International Corp Supplier No. P202411000.0
Each capsule contains 1g Peptone. Convenient and easy to use - simply add required number of premeasured capsules according to formulation being prepared without weighing and measuring.