Learn More
Alkali Scientific Peptone, 1 Gram Capsules, 500 G
SDP
Catalog No. 5000660289
Encompass_Preferred
Peptone, available in 1g capsules (500g total per bottle), is a premium protein supplement used to support the growth of a wide variety of microorganisms. This high-quality peptone provides essential amino acids and peptides that are critical for microbial metabolism. The pre-measured 1-gram capsules allow for easy and accurate preparation of culture media without the need for complex weighing or measuring. It is ideal for enriching microbiological media, enhancing microbial growth, and ensuring consistent results in bacterial culture, fermentation, and other microbiological applications. Peptone is commonly used in both research laboratories and industrial microbiological processes.
Catalog No. 50-006-60289 Supplier Alkali Scientific Supplier No. EPT002CP
Peptone, available in 1g capsules (500g total per bottle), is a premium protein supplement used to support the growth of a wide variety of microorganisms. This high-quality peptone provides essential amino acids and peptides that are critical for microbial metabolism. The pre-measured 1-gram capsules allow for easy and accurate preparation of culture media without the need for complex weighing or measuring. It is ideal for enriching microbiological media, enhancing microbial growth, and ensuring consistent results in bacterial culture, fermentation, and other microbiological applications. Peptone is commonly used in both research laboratories and industrial microbiological processes.