Learn More
Crescent Chemical Co Inc PEPTONE FROM CASEIN ENZYMATIC
SDP
Catalog No. 50249156
Encompass_Preferred
Peptone from casein enzymatic High quality source of peptides and amino acids produced by enzymatic digestion of casein. Refined hydrolysate that has been specially processed to increase solubility. Suitable as nutrient for laboratory media and industrial fermentation.
Catalog No. 50-249-156 Supplier Crescent Chemical Co Inc Supplier No. SE48600.02
Peptone from casein enzymatic High quality source of peptides and amino acids produced by enzymatic digestion of casein. Refined hydrolysate that has been specially processed to increase solubility. Suitable as nutrient for laboratory media and industrial fermentation.