Learn More
Sigma Aldrich Fine Chemicals Biosciences Peptone from soybean, enzymatic digest, 91079-46-8, MFCD07371265, 500 g
Catalog No. NC0793743
Encompass
For microbiology, Synonym: Peptone from soybean. This peptone is obtained by papaic digestion of soya flour. In additon to its nitrogen constituents, this peptone has a high carbohydrate content and is suitable for many purposes. It is widely used in culture media and is often used for the cultivation of many fastidious organisms and where rapid, luxuriant growth is required. However, due to this high content of sugar it is not recommendable for fermentation assays.
Catalog No. NC0793743 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 70178500G
For microbiology, Synonym: Peptone from soybean. This peptone is obtained by papaic digestion of soya flour. In additon to its nitrogen constituents, this peptone has a high carbohydrate content and is suitable for many purposes. It is widely used in culture media and is often used for the cultivation of many fastidious organisms and where rapid, luxuriant growth is required. However, due to this high content of sugar it is not recommendable for fermentation assays.