Learn More
Research Products International Corp Peptone, Granulated, 1 Kilogram
SDP
Catalog No. 50488898
Encompass_Preferred
Mixtures of peptides and free amino acids from peptic digest of animal tissue.
  • Amino Nitrogen (AN): 2.5-3.5%
  • Loss on Drying: <6.0%
Catalog No. 50-488-898 Supplier Research Products International Corp Supplier No. P202501000.0
Mixtures of peptides and free amino acids from peptic digest of animal tissue.
  • Amino Nitrogen (AN): 2.5-3.5%
  • Loss on Drying: <6.0%